Skip to Content
Merck
All Photos(6)

Documents

HPA018125

Sigma-Aldrich

Anti-SMPD2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Lyso-PAF-PLC, Anti-Lyso-platelet-activating factor-phospholipase C, Anti-N-SMase, Anti-Neutral sphingomyelinase, Anti-Sphingomyelin phosphodiesterase 2, Anti-nSMase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

QFIHHTSKKADVVLLCGDLNMHPEDLGCCLLKEWTGLHDAYLETRDFKGSEEGNTMVPKNCYVSQQELKPFPFGVRIDYVLYKAVSGFYISCKSFETTTGFDPHRGTPLSDHEALMATLFVRHSPPQQNPSST

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMPD2(6610)

General description

Sphingomyelin phosphodiesterase 2 (SMPD2) is present in the endoplasmic reticulum and Golgi apparatus.

Immunogen

Sphingomyelin phosphodiesterase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Sphingomyelin phosphodiesterase 2 (SMPD2) is involved in hydrolysing sphingomyelin and production of ceramide. Studies have shown that recombinant SMPD2 induces vascular smooth muscle cell (VSMC) apoptosis which is essential for the advancement of vascular calcification. In gonococcal infections, it takes part in recruiting phosphoinositide-3 kinase (PI3 kinase) to caveolin and thereby induces PI3 kinase-dependent downstream pathways. It also functions in activating cellular responses to hyperosmolar stress and contributes to the generation of ceramide, which is involved in stress-mediated apoptosis. SMPD2 takes part in the regulation of lipid droplet formation and cytokine secretion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73060

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Faulstich et al.
Cellular microbiology, 17(2), 241-253 (2014-09-17)
Disseminated gonococcal infection (DGI) is a rare but serious complication caused by the spread of Neisseria gonorrhoeae in the human host. Gonococci associated with DGI mainly express the outer membrane protein PorBIA that binds to the scavenger receptor expressed on
T Yabu et al.
Cell death and differentiation, 22(2), 258-273 (2014-08-30)
Neutral sphingomyelinase (nSMase) activation in response to environmental stress or inflammatory cytokine stimuli generates the second messenger ceramide, which mediates the stress-induced apoptosis. However, the signaling pathways and activation mechanism underlying this process have yet to be elucidated. Here we
Lizhen Liao et al.
PloS one, 8(12), e82379-e82379 (2013-12-21)
Vascular calcification is associated with significant cardiovascular morbidity and mortality, and has been demonstrated as an actively regulated process resembling bone formation. Oxidized low density lipoprotein (Ox-LDL) has been identified as a regulatory factor involved in calcification of vascular smooth
Alexandra Robciuc et al.
Journal of lipid research, 53(11), 2286-2295 (2012-08-18)
Hyperosmolarity (HO) imposes a remarkable stress on membranes, especially in tissues in direct contact with the external environment. Our efforts were focused on revealing stress-induced lipid changes that precede the inflammatory cytokine response in human corneal epithelial cells exposed to
Kerstin Menck et al.
Journal of extracellular vesicles, 6(1), 1378056-1378056 (2017-12-01)
Extracellular vesicles (EVs) are membrane particles secreted from cells into all body fluids. Several EV populations exist differing in size and cellular origin. Using differential centrifugation EVs pelleting at 14,000 g ("microvesicles" (MV)) and 100,000 g ("exosomes") are distinguishable by protein markers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service