Skip to Content
Merck
All Photos(2)

Key Documents

HPA013949

Sigma-Aldrich

Anti-MGP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell growth-inhibiting gene 36 protein, Anti-MGP, Anti-Matrix Gla protein precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

ESMESYELNPFINRRNANTFISPQQRWRAKVQERIRERSKPVHELNREACDDYRLCERYAMVYGYNAAYNRYFR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MGP(4256)

General description

Matrix-Gla Protein is a member of the Gla protein family and is found to be present in body fluids and locally present in osseous and extra-osseous tissue. The expression of this protein is abundant in renal tubular epithelia. The protein comprises of 84-aa mature protein and a 19-aa transmembrane signal peptide. It contains five to six amino acid residues that are vitamin K-dependent and γ-carboxyglutamic acid (Gla). The molar mass is found to be 15,000Da. The gene MGP is mapped to human chromosome 12p12.3.

Immunogen

Matrix Gla protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-MGP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The gene MGP (Matrix-Gla Protein) encodes an anti-calcifying factor that regulates the calcium crystal deposition in bone and teeth. It also functions in the inhibition of unwanted calcification in extra-osseous tissue such as renal cells. The activity of this protein is dependent on vitamin K. Alterations in the expression of this gene may result in increased vascular calcification and formation of urinary stones. Mutation in this gene has been associated with Keutel syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72585

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Price et al.
Biochemical and biophysical research communications, 117(3), 765-771 (1983-12-28)
A new protein has been isolated from CaCl2/urea extracts of demineralized bovine bone matrix. This protein has five to six residues of the vitamin K-dependent amino acid, gamma-carboxyglutamic acid (Gla), and we have accordingly designated it matrix Gla protein. Matrix
Johan M Lorenzen et al.
PloS one, 7(12), e52039-e52039 (2013-01-04)
Calcification of renal allografts is common in the first year after transplantation and is related to hyperparathyroidism. It is associated with an impaired long-term function of the graft (Am J Transplant 5∶1934-41, 2005). Aim of this study is to examine
P B Munroe et al.
Nature genetics, 21(1), 142-144 (1999-01-23)
Keutel syndrome (KS, MIM 245150) is an autosomal recessive disorder characterized by abnormal cartilage calcification, peripheral pulmonary stenosis and midfacial hypoplasia. A genome search using homozygosity mapping provided evidence of linkage to chromosome 12p12.3-13.1 (maximum multipoint lod score, 4.06). MGP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service