Skip to Content
Merck
All Photos(7)

Key Documents

HPA003600

Sigma-Aldrich

Anti-MAPRE1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APC-binding protein EB1 antibody produced in rabbit, Anti-EB1 antibody produced in rabbit, Anti-End-binding protein 1 antibody produced in rabbit, Anti-Microtubule-associated protein RP/EB family member 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
rat, mouse, human
citations:
5
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VAPSLVAPALNKPKKPLTSSSAAPQRPISTQRTAAAPKAGPGVVRKNPGVGNGDDEAAELMQQVNVLKLTVEDLEKERDFYFGKLRNIELICQENEGENDPVLQRIVDILYATDEGFVIPDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPRE1(22919)

General description

Microtubule-associated protein RP/EB family member 1 is a protein encoded by the MAPRE1 gene in humans. It is referred to as EB1 and encodes a 268-amino acid protein. The gene is mapped to human chromosome 20q11.

Immunogen

Microtubule-associated protein RP/EB family member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Microtubule-associated protein RP/EB family member 1 is a novel component of the microtubule cytoskeleton in mammalian cells. In association with mitotic apparatus, it may play a physiologic role connecting APC to cellular division, coordinating the control of normal growth and differentiation processes in the colonic epithelium. It also interacts with the plus-ends of microtubules and targets APC to microtubule tips. The interaction between this gene and APC plays an essential role in the regulation of microtubule polymerization. It acts as a potential biomarker and therapeutic target in CRC.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86553

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L K Su et al.
Cancer research, 55(14), 2972-2977 (1995-07-15)
Mutations of the APC gene play a critical role in both sporadic and familial forms of colorectal cancer. The vast majority of these mutations result in the loss of the carboxyl terminus of the protein. To further elucidate the function
M Nakamura et al.
Current biology : CB, 11(13), 1062-1067 (2001-07-27)
Human EB1 was originally cloned as a protein that interacts with the COOH terminus of adenomatous polyposis coli (APC). Interestingly, this interaction is often disrupted in colon cancer, due to mutations in APC. EB1 also interacts with the plus-ends of
Xin Dong et al.
The Journal of pathology, 220(3), 361-369 (2009-12-08)
Microtubule end-binding protein 1 (EB1) is an evolutionarily conserved protein that regulates microtubule dynamics and participates in diverse cell activities. Here, we demonstrate that EB1 expression is up-regulated in human breast cancer specimens and cell lines. The level of EB1
L Berrueta et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(18), 10596-10601 (1998-09-02)
The evolutionarily conserved protein EB1 originally was identified by its physical association with the carboxyl-terminal portion of the adenomatous polyposis coli (APC) tumor suppressor protein, an APC domain commonly mutated in familial and sporadic forms of colorectal neoplasia. The subcellular
Yutaka Sugihara et al.
Journal of proteomics, 75(17), 5342-5355 (2012-06-28)
Novel candidates of biomarker and therapeutic target in colorectal cancer (CRC) were investigated using a proteomic approach. The proteome of normal colorectal epithelial tissues was compared with that of the tumor ones in 59 CRC patients using two-dimensional difference gel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service