H3040
HER2 (676-end), active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
ERBB2, NEU, NGL, TKR1, c-erb B2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
9-13 nmol/min·mg
mol wt
~116 kDa
UniProt accession no.
relevant disease(s)
cancer (breast)
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... ERBB2(2064)
General description
HER2 (eceptor tyrosine-protein kinase erbB-2) is a cell surface-associated glycoprotein tyrosine kinase receptor.
Biochem/physiol Actions
HER2 (receptor tyrosine-protein kinase erbB-2) is an oncogene and is up-regulated in breast cancer. Overexpression of HER2 is associated with uncontrolled cellular proliferation, resistance to apoptotic cell death and increased motility.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R M Hudziak et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(20), 7159-7163 (1987-10-01)
The HER2 gene encodes a cell-surface glycoprotein with extensive homology to the epidermal growth factor receptor. Recently it was found to be amplified in about 30% of primary human breast malignancies. In experiments designed to assess the role of the
Ali Badache et al.
Journal of mammary gland biology and neoplasia, 11(1), 13-25 (2006-09-02)
Overexpression of the ErbB2/Her2 receptor tyrosine kinase in breast cancers is associated with the most aggressive tumors. Experimental studies have revealed that ErbB2 shows many features of a therapeutic target: ErbB2 is able to confer many of the characteristics of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service