Skip to Content
Merck
All Photos(1)

Key Documents

EHU046081

Sigma-Aldrich

MISSION® esiRNA

targeting human FAT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AATTGCTGACAACGCCTCTCCGAAGTTTACATCAAAAGAATATTCTGTTGAACTTAGTGAAACTGTCAGCATTGGGAGTTTCGTTGGGATGGTTACAGCCCATAGTCAATCATCAGTGGTGTATGAAATAAAAGATGGAAATACAGGTGATGCTTTTGATATTAATCCACATTCTGGAACTATCATCACTCAGAAAGCCCTGGACTTTGAAACTTTGCCCATTTACACATTGATAATACAAGGAACTAACATGGCTGGTTTGTCCACTAATACAACGGTTCTAGTTCACTTGCAGGATGAGAATGACAACGCGCCAGTTTTTATGCAGGCAGAATATACAGGACTCATTAGTGAATCAGCCTCAATTAACAGCGTGGTCCTAACAGACAGGAATGTCCCACTGG

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evanka Madan et al.
International journal of cancer, 139(11), 2570-2582 (2016-08-19)
The hypoxic microenvironment is an important contributor of glioblastoma (GBM) aggressiveness via HIF1α, while tumour inflammation is profoundly influenced by FAT Atypical Cadherin (FAT1). This study was designed to explore the functional interaction and significance of FAT1 and HIF1α under
Andrey Sheyko et al.
Nature, 539(7630), 551-554 (2016-11-08)
A striking feature of many natural dynamos is their ability to undergo polarity reversals. The best documented example is Earth's magnetic field, which has reversed hundreds of times during its history. The origin of geomagnetic polarity reversals lies in a
Xinhui Wu et al.
Cell biology international, 41(1), 24-32 (2016-10-21)
Porcine cumulus cells are localized around oocytes and act as a specific type of granulosa that plays essential roles in the development and maturation of oocytes, the development and atresia of follicles, and the development of embryos. Studies of FAT1
Tung-Nien Hsu et al.
Cancers, 11(12) (2019-12-01)
FAT atypical cadherin 1 (FAT1) regulates cell-cell adhesion and extracellular matrix architecture, while acting as tumor suppressor or oncogene, context-dependently. Despite implication of FAT1 in several malignancies, its role in oral squamous cell carcinoma (OSCC) remains unclear. Herein, we document
Longyue L Cao et al.
Nature, 539(7630), 575-578 (2016-11-10)
Mitochondrial products such as ATP, reactive oxygen species, and aspartate are key regulators of cellular metabolism and growth. Abnormal mitochondrial function compromises integrated growth-related processes such as development and tissue repair, as well as homeostatic mechanisms that counteract ageing and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service