CLS70581500
Pyrex® VISTA™ volumetric flask, Class B
capacity 500 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
long
sterility
non-sterile
feature
closure type Snap cap
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 70581-500
capacity
500 mL
Related Categories
General description
These Pyrex VISTA Class B volumetric flasks have capacity tolerances that are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue clock letters make inscriptions easy to read. Snap-caps are supplied with all sizes.
Legal Information
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)