Skip to Content
Merck
All Photos(2)

Key Documents

CLS356486

Corning® BioCoat Collagen Cell Culture Flask

capacity 150 cm2, rectangular flask, canted neck, vented, Cap

Synonym(s):

cell culture flask, plastic cell culture flask, coated flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

Pricing and availability is not currently available. Contact Technical Service

material

rectangular flask

feature

vented Cap
canted neck

packaging

pkg of 40 ea (8 packs of 40)

manufacturer/tradename

Corning 356486

capacity

150 cm2

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service