Skip to Content
Merck
All Photos(1)

Key Documents

CLS3477

Corning® NETWELL® 12 well plates

polyester membrane, round wells, clear wells, clear plate, polyester plate, sterile, pkg of 48 (4 x 12 well inserts)

Synonym(s):

cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

Product Name

Corning® Netwell® inserts, membrane diam. 15 mm, membrane size 74 μm , mesh

material

clear plate
clear wells
polyester membrane
polyester plate
round wells

sterility

sterile

feature

lid
skirt

packaging

pkg of 48 (4 x 12 well inserts)

manufacturer/tradename

Corning 3477

membrane diam.

15 mm

membrane size

74 μm , mesh

size

12 wells , per insert

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Netwells are polystyrene inserts fitted with polyester mesh bottoms that function as tissue carriers and strainers for:
  • Immunocytochemical staining
  • Explant culture
  • 3-D tissue culture
  • Coarse filtration of cell suspension

Features and Benefits

  • Corning® Netwell® inserts are 15mm diameter netwell inserts with 74μm polyester mesh bottoms attached to polystyrene rings or housing.
  • Supplied as sterile and pre-loaded in 12-well microplates.

Legal Information

Corning is a registered trademark of Corning, Inc.
NETWELL is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service