Skip to Content
Merck
All Photos(2)

Key Documents

CLS3232

Corning® CellCube®

25 layer, TCT, with 6” Silicone tubing and Aseptic Connectors, sterile, bag of 1 ea, case of 1 ea

Synonym(s):

cell culture vessel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

polystyrene
silicon tubing (6′′)

sterility

sterile

packaging

bag of 1 ea
case of 1 ea

manufacturer/tradename

Corning 3232

surface area

21,250 cm2 , cell growth area

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Introducing Corning′s Closed System Cell Cube – a new closed system offering designed to help reduce the risk of adventitious contamination. Our new offering of CellCube 10, 25, 100 modules, circulation loops, and connectors allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik® and MPC connectors.

Application

Adherent Cell Scale Up

Features and Benefits

  • 21,250 cm2 Surface Area
  • Corning TCT surface treatment
  • Sterility Assurance Level(SAL) of 10-6
  • AseptiQuik® G Connector

Legal Information

AseptiQuik is a registered trademark of Colder Products Company
CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service