Skip to Content
Merck
All Photos(1)

Key Documents

HT55 Cell Line human

85061105, human colon, Epithelial

Synonym(s):

HT55 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human colon
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

HT55 Cell Line human, 85061105

biological source

human colon

description

Human colon carcinoma

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon carcinoma

Cell Line Description

Human colon epithelial cell line

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 9,13
D16S539: 14
D5S818: 11,12
D7S820: 11,13
THO1: 8,9.3
TPOX: 8,9
vWA: 14,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA and incubate until cell sheet peels off. Cells grow in islands, not monolayers; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service