BAF189050051
Centrifuge Tube Ice Rack
Rack Cover Only, polypropylene
Synonym(s):
cold tube storage
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122806
NACRES:
NB.22
Recommended Products
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18905-0051
technique(s)
cryopreservation: suitable
L × W
25.4 cm (9⅛ in.) × 13.3 cm (5¼ in.)
Looking for similar products? Visit Product Comparison Guide
General description
Large Spill Containment Tray Conveniently capture fluid spills when decanting or pouring hazardous or toxic liquids from one container to another. The unit size is ideal for capturing moderately sized spills on the benchtop or in the field. Several flasks, beakers or a rack of test tubes can be placed on the spill support. Hazardous, corrosive substances, dyes or stains may also be stored in the tray.
- Dual convenience: Bench top ice rack and Large Spill containment system
- Durable, stackable ice rack holds approximately 67 oz (2 liters) of ice
- The large chilled bath holds twenty-eight 50 ml centrifuge tubes
- Durable polypropylene construction offers chemical resistance
- Steam sterilizable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service