Skip to Content
Merck
All Photos(3)

Key Documents

AV37583

Sigma-Aldrich

Anti-E2F7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-A630014C11Rik, Anti-D10Ertd739e, Anti-E2F transcription factor 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

99 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... E2f7(52679)

General description

E2F transcription factors are key regulators of cell growth, differentiation and cell death. E2F transcription factor 7 (E2F7) which is highly expressed during mid to late S-phase binds to the promoter regions of and represses G(1)/S-regulated genes thus promoting the downswing of oscillating G(1)/S genes during S-phase progression.

The previously assigned protein identifier Q8BSQ3 has been merged into Q6S7F2. Full details can be found on the UniProt database.

Specificity

Anti-E2F7 polyclonal antibody reacts with human, rat, canine, bovine, and mouse E2F transcription factor 7 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of mouse E2f7

Application

Anti-E2F7 polyclonal antibody is used to tag E2F transcription factor 7 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of E2F transcription factor 7 in the regulation of cell proliferation, especially at the level of G(1)/S gene activity during S-phase progression.

Biochem/physiol Actions

E2F7 is a E2F family member. It can block the E2F-dependent activation of a subset of E2F target genes as well as mitigate cellular proliferation of mouse embryo fibroblasts

Sequence

Synthetic peptide located within the following region: MEVNCLTLKDLISPRQTRLDFAIEDAENAQKENIFVDRSRMTPKTPMKNE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhongkui Jin et al.
Hereditas, 161(1), 27-27 (2024-08-28)
Circular RNAs (circRNAs) are capable of affecting breast cancer (BC) development. However, the role and underneath mechanism of circFKBP8 (also known as hsa_circ_0000915) in BC remain largely unknown. Expression analyses were performed using quantitative real-time polymerase chain reaction (qRT-PCR), western
Marie-Claude Perry et al.
Molecular and cellular biology, 34(23), 4232-4243 (2014-09-24)
The tyrosine kinase receptor ERBB2 is required for normal development of the heart and is a potent oncogene in breast epithelium. Trastuzumab, a monoclonal antibody targeting ERBB2, improves the survival of breast cancer patients, but cardiac dysfunction is a major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service