Skip to Content
Merck
All Photos(1)

Documents

A1878

Sigma-Aldrich

Ala-Leu

≥99% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O3
CAS Number:
Molecular Weight:
202.25
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Ala-Leu,

Assay

≥99% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)N)C(O)=O

InChI

1S/C9H18N2O3/c1-5(2)4-7(9(13)14)11-8(12)6(3)10/h5-7H,4,10H2,1-3H3,(H,11,12)(H,13,14)/t6-,7-/m0/s1

InChI key

RDIKFPRVLJLMER-BQBZGAKWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Alanyl dipeptides such as ala-leu, ala-lys, ala-gly, ala-pro, ala-tyr and ala-phe may be used in physicochemical studies or to evaluate dipeptide separation technologies. Alanyl dipeptides may also be used for studying cell uptake mechanisms, dipeptide metabolism or cell growth supplementation benefits.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew Rosati et al.
Molecular and biochemical parasitology, 217, 7-12 (2017-08-16)
The aminopeptidase PfA-M1 is a key contributor to peptide catabolism in the human malaria parasite Plasmodium falciparum. PfA-M1 substrate specificity is shaped by the cylindrical S1 subsite, which accommodates the sidechain of the substrate P1 residue. At the top of
Yafang Tu et al.
Molecular immunology, 121, 59-71 (2020-03-13)
Tristetraprolin (TTP) regulates inflammatory and immune responses by destabilizing target mRNAs via binding to their 3'-UTR AREs. We have recently reported that TTP preferentially up-regulates the expression level of innate immunity genes involved in the type I interferon-mediated signaling pathway
Alice B Nongonierma et al.
Food research international (Ottawa, Ont.), 94, 79-89 (2017-03-16)
The release of dipeptidyl peptidase IV (DPP-IV) inhibitory peptides from bovine milk protein isolate (MPI) during trypsin hydrolysis was studied using a design of experiments (DOE) approach. A 3 factor×3 level DOE including temperature (40, 50 and 60°C), enzyme to
Víctor Cruz et al.
The journal of physical chemistry. B, 115(16), 4880-4886 (2011-04-08)
An exhaustive umbrella sampling simulation of the alanine dipeptide in solution is reported. The presence of stable Y conformations in solution is assessed by both quantum calculations and experimental data from X-ray and NMR protein-solved structures available in the protein
Trang U Vo et al.
Electrophoresis, 25(9), 1230-1236 (2004-06-03)
Series of dipeptides, including homodipeptides and alanyl dipeptides, were separated using quadruplex (G-quartet) DNA stationary phases in open-tubular capillary electrochromatography (OTCEC). The stationary phases were constructed by covalently attaching the DNA oligonucleotides to the inner capillary surface. Three different G-quartet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service