Skip to Content
Merck
All Photos(1)

Key Documents

56189

Sigma-Aldrich

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester

≥97.0% (C), BioReagent

Synonym(s):

N-Succinimidyl 3-(4-hydroxyphenyl)propionate, N-[(4-Hydroxyhydrocinnamoyl)oxy]succinimide, Bolton-Hunter reagent precursor, Rudinger reagent, SHPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO5
CAS Number:
Molecular Weight:
263.25
Beilstein:
1545011
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

product name

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester, suitable for fluorescence, BioReagent, ≥97.0% (C)

product line

BioReagent

Quality Level

Assay

≥97.0% (C)

mp

132-133 °C (lit.)

fluorescence

λex 320 nm; λem 409 nm in 0.1 M Tris pH 8.8, 2 mM H2O2, peroxidase

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

Oc1ccc(CCC(=O)ON2C(=O)CCC2=O)cc1

InChI

1S/C13H13NO5/c15-10-4-1-9(2-5-10)3-8-13(18)19-14-11(16)6-7-12(14)17/h1-2,4-5,15H,3,6-8H2

InChI key

KYRUKRFVOACELK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bolton-Hunter reagent precursor (3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester) is a hydroxysuccinimide ester used as an intermediate for high radioactive radioiodination of proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D P McManus et al.
Journal of helminthology, 61(1), 47-52 (1987-03-01)
Living, intact protoscoleces of the British horse and sheep strains of Echinococcus granulosus were subjected to surface radioiodination procedures using 125I and Iodogen and 125I-Bolton Hunter reagent. Subsequent combined electron microscopy and autoradiography revealed specific surface membrane labelling with the
Jae Jun Park et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(1), 56-62 (2010-10-05)
In this study, the radioiodination and biodistribution of quantum dots (QDs) using Bolton-Hunter reagent were investigated. Radioiodination yield was 33.4 ± 2.0%. Fluorescent intensity of radioiodinated QDs decreased to 75.4% of the maximum prior to radioiodination. In biodistribution and ex
Appendix: Preparation of N-succinimidyl 3-(4-hydroxyphenyl)propionate.
J Rudinger et al.
The Biochemical journal, 133(3), 538-539 (1973-07-01)
M Rack et al.
Pflugers Archiv : European journal of physiology, 400(4), 403-408 (1984-04-01)
Several reagents that react with sulfhydryl and amino groups were applied to voltage-clamped single nerve fibres of the frog. The fibres were exposed to comparable amounts of the chemical reagents for relatively short times. 3-(p-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester (HPPS), a
Radioiodination of cellular proteins.
Caplan S, Baniyash M.
Current Protocols in Cell Biology, 7, 7-7 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service