Skip to Content
Merck
All Photos(1)

Documents

QC3077

Supelco

Dissolved Oxygen - Whole Volume

certified reference material

Synonym(s):

Whole-Volume Dissolved Oxygen Measurement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-3263

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application


  • Cysteamine hydrochloride in lysosomal storage disease research: Cysteamine hydrochloride is utilized in treatments for lysosomal storage diseases, such as cystinosis. Its effectiveness in reducing cystine levels in cells highlights its critical role in biochemical studies focused on metabolic dysfunctions (Sun C et al., 2024).

  • Cysteamine HCl in Huntington′s disease therapeutic research: Research on cysteamine hydrochloride has explored its use in models of Huntington′s disease, where its properties may alleviate symptoms or modify disease progression. This aligns with its broader use in neurodegenerative disease research, showcasing its versatility and therapeutic potential (Montaseri Z et al., 2023).

  • Cysteamine hydrochloride as a transglutaminase inhibitor: The application of cysteamine hydrochloride extends to its role as a transglutaminase inhibitor, which is significant in studies related to cellular functions and diseases where transglutaminase activity is a factor, furthering our understanding of its biochemical interactions and therapeutic implications (Aliyu AW et al., 2023).


Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service