NIST495
Unalloyed copper
NIST® SRM® 495, Cu II (solid)
Synonym(s):
Copper
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
grade
certified reference material
Quality Level
packaging
pkg of rod
manufacturer/tradename
NIST®
resistivity
1.673 μΩ-cm, 20°C
bp
2567 °C (lit.)
mp
1083.4 °C (lit.)
density
8.94 g/mL at 25 °C (lit.)
format
matrix material
SMILES string
[Cu]
InChI
1S/Cu
InChI key
RYGMFSIKBFXOCR-UHFFFAOYSA-N
General description
Other Notes
Certified for the analytes listed below.
Antimony (Sb), Arsenic (As), Bismuth (Bi), Chromium (Cr), Lead (Pb), Manganese (Mn), Nickel (Ni), Selenium (Se), Silver (Ag), Sulfur (S), Tellurium (Te), Tin (Sn), Zinc (Zn), Copper (Cu)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Cadmium (Cd), Gold (Au), Iron (Fe), Magnesium (Mg), Oxygen (O), Silicon (Si), Calcium (Ca), Titanium (Ti)
See certificate for values and more details at nist.gov/SRM.
Antimony (Sb), Arsenic (As), Bismuth (Bi), Chromium (Cr), Lead (Pb), Manganese (Mn), Nickel (Ni), Selenium (Se), Silver (Ag), Sulfur (S), Tellurium (Te), Tin (Sn), Zinc (Zn), Copper (Cu)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Cadmium (Cd), Gold (Au), Iron (Fe), Magnesium (Mg), Oxygen (O), Silicon (Si), Calcium (Ca), Titanium (Ti)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service