E1903
Ethidium homodimer I solution
≥95% purity (HPLC), liquid
Synonym(s):
EtDi, EthD-1, Phenanthridinium, 5,5′-[1,2-ethanediylbis(imino-3,1-propanediyl)]bis(3, 8-diamino-6-phenyl)-,dichloride,dihydrochloride
About This Item
Recommended Products
Product Name
Ethidium homodimer I solution, ≥95% (Run on Ethidium Homodimer before dissolution in DMSO, TLC), 2 mM in DMSO
Quality Level
Assay
≥95% (Run on Ethidium Homodimer before dissolution in DMSO, TLC)
form
liquid
concentration
2 mM in DMSO
technique(s)
titration: suitable
color
red
ε (extinction coefficient)
≥9000 at 490-496 nm in water
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
−20°C
SMILES string
Cl.Cl.[Cl-].[Cl-].Nc1ccc2c(c1)c(-c3ccccc3)[n+](CCCNCCNCCC[n+]4c(-c5ccccc5)c6cc(N)ccc6c7ccc(N)cc47)c8cc(N)ccc28
InChI
1S/C46H46N8.4ClH/c47-33-13-17-37-39-19-15-35(49)29-43(39)53(45(41(37)27-33)31-9-3-1-4-10-31)25-7-21-51-23-24-52-22-8-26-54-44-30-36(50)16-20-40(44)38-18-14-34(48)28-42(38)46(54)32-11-5-2-6-12-32;;;;/h1-6,9-20,27-30,49-52H,7-8,21-26,47-48H2;4*1H
InChI key
ZKAJUKABUUJRLB-UHFFFAOYSA-N
Application
Biochem/physiol Actions
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
185.0 °F - closed cup
Flash Point(C)
85 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service