Skip to Content
Merck
All Photos(1)

Key Documents

80330

Millipore

Enrichment ChromoSelect Broth Base for EC O157:H7

NutriSelect® Plus, suitable for microbiology

Synonym(s):

EC O157:H7 ChromoSelect Enrichment Broth Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bile salts mixture, 1.5 g/L
casein enzymic hydrolysate, 10.0 g/L
chromogenic mixture, 1.3 g/L
sorbitol, 10.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.1±0.2 (25 °C)

application(s)

agriculture
food and beverages

microbiology

storage temp.

2-8°C

suitability

selective and differential for enterovirulent E. coli (EEC)

Application

HiCrome Enrichment Broth (Base) for EC 0157:H7 is recommended for isolation and selective differentiation of E. coli 0157:H7 from food and environmental samples by chromogenic method.

Preparation Note

Suspend 11.4 g in 495 ml distilled water. Heat to boiling to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 45-50°C and aseptically add the contents of 1 vial of EC 0157:H7 ChromoSelect Selective Supplement I (Cat. No. 80331) to the medium. Mix well and pour into sterile test tubes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
HiCrome is a trademark of HiMedia Laboratories Pvt. Ltd.
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service