Skip to Content
Merck
All Photos(2)

Key Documents

76773

Supelco

Victoria Pure Blue BO

analytical standard

Synonym(s):

Basic Blue 7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40ClN3
CAS Number:
Molecular Weight:
514.14
Colour Index Number:
42595
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

[Cl-].CCNc1ccc(\C(c2ccc(cc2)N(CC)CC)=C3/C=C\C(C=C3)=[N+](\CC)CC)c4ccccc14

InChI

1S/C33H39N3.ClH/c1-6-34-32-24-23-31(29-13-11-12-14-30(29)32)33(25-15-19-27(20-16-25)35(7-2)8-3)26-17-21-28(22-18-26)36(9-4)10-5;/h11-24H,6-10H2,1-5H3;1H

InChI key

ROVRRJSRRSGUOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Victoria Pure Blue BO (VBPO) is a cationic dye belonging to the triarylmethane family. Victoria Pure Blue BO-deethyl metabolite is reported as a potential biomarker for controlling the content of VPBO in farmed fish foodstuffs.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Victoria Pure Blue BO may be used as an ion pairing reagent in:
  • Determination of molybdenum (VI) in food samples and beverages using flame atomic absorption spectrometry (FAAS).
  • Determination of inorganic antimony species in beverages and biological samples beverages using flame atomic absorption spectrometry (FAAS).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro investigations of the metabolism of Victoria pure blue BO dye to identify main metabolites for food control in fish
Dubreil E, et al.
Chemosphere, 238 (2020)
A simple and efficient approach for preconcentration of some heavy metals in cosmetic products before their determinations by flame atomic absorption spectrometry
Altunay N and Gurkan R
Turkish Journal of Chemistry, 40(6), 988-1001 (2016)
Determination of low levels of molybdenum (VI) in food samples and beverages by cloud point extraction coupled with flame atomic absorption spectrometry
Gurkan R, et al.
J. Food Compos. Anal., 32(1), 74-82 (2013)
A new cloud point extraction procedure for determination of inorganic antimony species in beverages and biological samples by flame atomic absorption spectrometry
Altunay N and Gurkan R
Food Chemistry, 175, 507-515 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service