Skip to Content
Merck
All Photos(1)

Documents

74332

Supelco

Diazinon-(diethyl-d10)

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D10H11N2O3PS
CAS Number:
Molecular Weight:
314.41
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥95 atom % D

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.4978 (lit.)

bp

83-84 °C/0.002 mmHg (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+10

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])OP(=S)(Oc1cc(C)nc(n1)C(C)C)OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C12H21N2O3PS/c1-6-15-18(19,16-7-2)17-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3/i1D3,2D3,6D2,7D2

InChI key

FHIVAFMUCKRCQO-HXOHQZFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diazinon-(diethyl-d10) is an isotope labeled analog of the organophosphate pesticide diazinon, wherein the diethoxy protons are replaced by deuterium.

Application

Diazinon-(diethyl-d10) may be used as an internal standard for the determination of diazinon in soybeans and Daphnia magna by various chromatography techniques.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-isotope dilution mass spectrometry method validation for target pesticides in soybeans
Huertas-Perez, JF, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 36(1), 96-108 (2019)
In vitro rat hepatic and intestinal metabolism of the organophosphate pesticides chlorpyrifos and diazinon
Poet TS, et al.
Toxicological Sciences, 72(2), 193-200 (2003)
Toxicokinetic Model Describing Bioconcentration and Biotransformation of Diazinon in Daphnia magna
Preuss TG, et al.
Environmental Science & Technology, 45(1), 4995-5002 (2011)
Accurate determination of selected pesticides in soya beans by liquid chromatography coupled to isotope dilution mass spectrometry
Perez JFH, et al.
Talanta, 137(2), 120-129 (2015)

Articles

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service