Skip to Content
Merck
All Photos(1)

Key Documents

74173

Millipore

Preston Broth Base

suitable for microbiology, NutriSelect® Prime

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

Agency

according to ISO 10272-1:2017

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 1.0 g/L
peptone, 10.0 g/L
peptose*, 10.0 g/L (*equivalent to enzymatic digest of animal tissues)
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Prime

final pH

7.4±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Campylobacter spp.

Application

For selective enrichment and cultivation of Campylobacter species . The composition and performance criteria of this medium are as per the specifications laid down in ISO 10272-1:2017.

Preparation Note

Suspend 25 grams in 945 ml of purified/distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15lbs pressure (121°C) for 15 minutes. Cool to 45-50°C and aseptically add 50 ml of sterile lysed horse blood and reconstituted contents of 2 vials of Preston Selective Supplement (Cat. No. 38390). Mix well and dispense into sterile tubes or flasks as desired.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service