71009-576BC
GDS Listeria spp. Tq
suitable for RT-PCR, for Listeria spp.
About This Item
Recommended Products
Agency
according to Immunomagnetic Separation (IMS)
meets requirements for AOAC® (PTM)
meets requirements for Health Canada
meets requirements for MicroVal
Quality Level
product line
Assurance GDS®
shelf life
6-12 mo.
feature
0.1 mL tubes
packaging
pkg of 576 tests
technique(s)
RT-PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
configured for BioControl Systems GDS Rotor-Gene
configured for BioControl Systems GDS
storage temp.
2-8°C
suitability
Listeria spp.
General description
Application
Features and Benefits
- Results in 24 hours for environmental samples and 32 hours for product samples
- Single-step enrichment reduces handling time
- Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and faster time to results
- Simplified sample prep does not require heating blocks or preparation of reagents
- Proprietary Rotor-Gene® Q cycler is a rotory-based cycler that ensures uniform heating and cooling of all samples
Components
- Listeria spp. Amplification Tubes Tq (0.1 mL)
- Listeria Concentration Reagent
- Listeria Resuspension Buffer Tq
- Wash Solution
NOTE: The 100 kit includes standard 0.2 mL amp tubes whereas the 576 kit includes smaller 0.1 mL amp tubes. The 576 kit requires additional equipment tools to process per the kit Directions for Use.
Compatibility
Testing Method
Legal Information
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service