Skip to Content
Merck
All Photos(1)

Key Documents

67954

Supelco

4-(Dimethylamino)benzoyl chloride

for HPLC derivatization, LiChropur, ≥99.0% (HPLC)

Synonym(s):

DMABC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4COCl
CAS Number:
Molecular Weight:
183.63
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NB.21

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)
99.0-101.0% (AT)

quality

LiChropur

technique(s)

HPLC: suitable

mp

145-149 °C (lit.)
146-150 °C

suitability

complies for LC-MS

SMILES string

CN(C)c1ccc(cc1)C(Cl)=O

InChI

1S/C9H10ClNO/c1-11(2)8-5-3-7(4-6-8)9(10)12/h3-6H,1-2H3

InChI key

UGJDXRVQCYBXAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Dimethylamino)benzoyl chloride is an organic chemical, basically used as HPLC derivatization reagent.
Material is reported to be very moisture sensitive. Handle under an inert atmosphere

Application

4-(Dimethylamino)benzoyl chloride dissovled in N-methyl-2-pyrrolidinone (NMP) and pyridine may be used in the synthesis of 6-O-(4-(Dimethylamino)benzoyl)curdlan (DABz-Cur) by adding curdlan to dry NMP.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gaku Fukuhara et al.
Chemical communications (Cambridge, England), 46(48), 9128-9130 (2010-11-10)
A newly synthesized chromophore-modified curdlan functions as a saccharide chemosensor in aqueous solution, enabling us to discriminate tetrasaccharide acarbose from 24 mono-, di-, tri-, and tetrasaccharides.
Yoshio Suzuki et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 20(3), 475-482 (2004-04-08)
Novel labeling reagents, called MS probes, which possess a positively charged quaternary amine moiety and can transform a neutral analyte into a charged compound by simply mixing with the analyte and allowing the mixture to stand from several minutes to

Articles

Improved LC/MS Detection of Estradiol using 4-(Dimethylamino) benzoyl chloride (DMABC) derivatization agent relative to dansyl chloride.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service