Skip to Content
Merck
All Photos(1)

Key Documents

41788

Supelco

Myristoleic acid

analytical standard

Synonym(s):

cis-9-Tetradecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
226.36
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4562 (lit.)

bp

144 °C/0.6 mmHg (lit.)

mp

−4.5-−4 °C (lit.)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C14H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h5-6H,2-4,7-13H2,1H3,(H,15,16)/b6-5-

InChI key

YWWVWXASSLXJHU-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Deena Bader et al.
Molecular and cellular biology (2020-12-09)
Lithium chloride (LiCl) is commonly used in treatment of mood disorders, however its usage leads to weight gain which promotes metabolic disorders. Protein Kinase C delta (PKCδ), a serine/threonine kinase, is alternatively spliced to PKCδI and PKCδII in 3T3L1 cells.
Dong-Hee Kang et al.
Nature communications, 12(1), 5059-5059 (2021-08-26)
With the current interest in cultured meat, mammalian cell-based meat has mostly been unstructured. There is thus still a high demand for artificial steak-like meat. We demonstrate in vitro construction of engineered steak-like tissue assembled of three types of bovine
Fátima Camões et al.
Biochimica et biophysica acta, 1853(1), 111-125 (2014-10-14)
Peroxisomes are ubiquitous organelles which participate in a variety of essential biochemical pathways. An intimate interrelationship between peroxisomes and mitochondria is emerging in mammals, where both organelles cooperate in fatty acid β-oxidation and cellular lipid homeostasis. As mitochondrial fatty acid
Guan-yuan Chen et al.
Clinica chimica acta; international journal of clinical chemistry, 438, 126-134 (2014-08-26)
As fatty acids play an important role in biological regulation, the profiling of fatty acid expression has been used to discover various disease markers and to understand disease mechanisms. This study developed an effective and accurate comparative fatty acid analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service