Skip to Content
Merck
All Photos(2)

Documents

33394

Supelco

MCPA-2-ethylhexyl ester

PESTANAL®, analytical standard

Synonym(s):

2-Ethylhexyl (4-chloro-2-methylphenoxy)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25ClO3
CAS Number:
Molecular Weight:
312.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

refractive index

n20/D (effective)

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCC(CC)COC(=O)COc1ccc(Cl)cc1C

InChI

1S/C17H25ClO3/c1-4-6-7-14(5-2)11-21-17(19)12-20-16-9-8-15(18)10-13(16)3/h8-10,14H,4-7,11-12H2,1-3H3

InChI key

IDGRPSMONFWWEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MCPA-2-ethylhexyl ester is a systemic, post-emergent herbicide used for the selective control of annual and perennial broad leaved weeds in cereals and other specific crops.

Application

MCPA-2-ethylhexyl ester may be used as an analytical standard for the determination of the analyte in formulations using high-performance liquid chromatography (HPLC) and gas chromatography (GC) as well as gas chromatography with mass spectrometric detection (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>NaN °F - Pensky-Martens closed cup

Flash Point(C)

> 133.3 - < 138.7 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phenoxy ester herbicides by gas and high-performance liquid chromatography.
Sanchez-Brunete C, et al.
Journal of Chromatography A, 552(4), 235-240 (1991)
Environmental fate and effects of MCPA: A Canadian perspective.
Caux YP, et al.
Critical reviews in environmental science and technology, 25(4), 313-376 (1995)
Re-evaluation of the Lawn and Turf Uses of the Herbicide (4-chloro-2-methylphenoxy) Acetic Acid (MCPA) (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service