Key Documents
20722
Hamilton® syringe, 1000 series GASTIGHT®, luer tip
1005LT, volume 5 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81501
needle L
, n/a
volume
5 mL
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)