Key Documents
20701
Hamilton® syringe, luer tip
705LT, volume 50 μL, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Pricing and availability is not currently available.
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80501
needle L
, n/a
volume
50 μL
compatibility
for use with Hamilton 705LT (Luer tip)
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Legal Information
Hamilton is a registered trademark of Hamilton Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Akhil Kotwal et al.
Cellular signalling, 76, 109801-109801 (2020-10-06)
Deregulated DNA methylation and post-translational histone modifications are majorly associated with cancer progression. Histone modification regulates the gene expression patterns that contribute to the emergence of sporadic cancers. Histone deacetylases (HDACs) act as erasers of acetylation marks, and their functions
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)