Skip to Content
Merck
All Photos(1)

Key Documents

14274

Supelco

Acetonitrile solution

with 0.1% ammonium acetate, tested for UHPLC-MS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NA.03
grade:
HPLC grade
technique(s):
HPLC: suitable

Product Name

Acetonitrile with 0.1% ammonium acetate, tested for UHPLC-MS

grade

HPLC grade

form

liquid

quality

tested for UHPLC-MS

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable

impurities

≤0.0001% non-volatile
≤0.5 ppb fluorescence (quinine) at 254 nm
≤0.5 ppb fluorescence (quinine) at 365 nm
≤2.0% water
≤20 ppb UHPLC-MS (q-TOF) (Reserpin, ESI(+))

transmittance

254 nm, ≥90%

cation traces

Ca: ≤0.25 mg/kg
K: ≤0.25 mg/kg
Mg: ≤0.25 mg/kg
Na: ≤1 mg/kg

UHPLC-gradient

≤2 mAU at 254 nm (peak)
≤30 mAU at 254 nm (drift)

suitability

passes test for UHPLC-MS

format

mixture

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetonitrile with ammonium acetate may be used as a mobile phase in liquid chromatography-tandem mass spectrometry (LC-MS/MS) assay for the simultaneous quantitative analysis of synthetic corticosteroids like beclomethasone dipropionate, betamethasone, budesonide, dexamethasone, fludrocortisone, flunisolide, fluorometholone, fluticasone propionate, megestrol acetate, methylprednisolone, prednisolone, prednisone, triamcinolone, and triamcinolone acetonide.

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantitative, highly sensitive liquid chromatography-tandem mass spectrometry method for detection of synthetic corticosteroids.
Taylor RL
Clinical Chemistry, 50 (12), 2345-2352 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service