Skip to Content
Merck
All Photos(1)

Key Documents

03880590

Luteolin

primary reference standard

Synonym(s):

3′,4′,5,7-Tetrahydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
Beilstein:
292084
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

mp

~330 °C (lit.)

application(s)

food and beverages

SMILES string

Oc1cc(O)c2C(=O)C=C(Oc2c1)c3ccc(O)c(O)c3

InChI

1S/C15H10O6/c16-8-4-11(19)15-12(20)6-13(21-14(15)5-8)7-1-2-9(17)10(18)3-7/h1-6,16-19H

InChI key

IQPNAANSBPBGFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: achillea

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Leticia Correa Manjolin et al.
Food chemistry, 141(3), 2253-2262 (2013-07-23)
Fisetin, quercetin, luteolin and 7,8-hydroxyflavone show high activity in Leishmania cultures and present low toxicity to mammalian cells. In this work, the structural aspects of 13 flavonoids were analyzed for their inhibition of the arginase enzyme from Leishmania (Leishmania) amazonensis.
Miguel López-Lázaro
Mini reviews in medicinal chemistry, 9(1), 31-59 (2009-01-20)
Epidemiological evidence suggests that flavonoids may play an important role in the decreased risk of chronic diseases associated with a diet rich in plant-derived foods. Flavonoids are also common constituents of plants used in traditional medicine to treat a wide
Benguo Liu et al.
Food chemistry, 141(2), 900-906 (2013-06-26)
The formation of supramolecular inclusion complexes between luteolin and five cyclodextrins namely β-cyclodextrin (β-CD), methyl-β-cyclodextrin (M-β-CD), hydroxyethyl-β-cyclodextrin (HE-β-CD), hydroxypropyl-β-cyclodextrin (HP-β-CD) and glucosyl-β-cyclodextrin (G-β-CD) was investigated. Results from phase-solubility studies showed that luteolin formed 1:1 stoichiometric inclusion complexes with these cyclodextrins
Chithan Kandaswami et al.
In vivo (Athens, Greece), 19(5), 895-909 (2005-08-16)
The flavonoids are polyphenolic compounds found as integral components of the human diet. They are universally present as constituents of flowering plants, particularly of food plants. The flavonoids are phenyl substituted chromones (benzopyran derivatives) consisting of a 15-carbon basic skeleton
Poyil Pratheeshkumar et al.
PloS one, 7(12), e52279-e52279 (2013-01-10)
Angiogenesis, the formation of new blood vessels from pre-existing vascular beds, is essential for tumor growth, invasion, and metastasis. Luteolin is a common dietary flavonoid found in fruits and vegetables. We studied the antiangiogenic activity of luteolin using in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service