Skip to Content
Merck
All Photos(5)

Key Documents

PFLRKB

Roche

KAPA PROBE FAST

suitable for qPCR, 2 ×, ROX Low

Synonym(s):

Probe qPCR, qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 100 reactions
sufficient for 1000 reactions
sufficient for 500 reactions

shelf life

≤12 mo.

feature

dNTPs included
hotstart

packaging

kit of 1 mL (100 x 20 μL rxn; KK4716)
kit of 10 mL (1000 x 20 μL rxn; KK4718)
kit of 5 mL (500 x 20 μL rxn; KK4717)
pkg of

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

qPCR: suitable

input

purified DNA

compatibility

for use with Agilent Mx3000P
for use with Agilent Mx3005P
for use with Agilent Mx4000
for use with Applied Biosystems® 7500
for use with Applied Biosystems® ViiA 7
for use with QuantStudio

detection method

probe-based

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA PROBE FAST qPCR Kits provide fast and reproducible results for all probe-based qPCR applications. Kits contain a ready-to-use master mix for highly sensitive and accurate real-time PCR using sequence-specific fluorogenic probe chemistries, including hydrolysis probes (e.g. TaqMan®), FRET probes, and displacement probes (e.g. molecular beacons).

Application

KAPA PROBE FAST qPCR Master Mix (2X) with ROX Low is suitable for use in:
  • Genotyping
  • Gene expression analysis
  • Multiplex qPCR
  • SNP genotyping/allelic discrimination
  • Microarray validation
  • Real time(RT)-quantitative polymerase chain reaction (qPCR)/qPCR

Features and Benefits

Accurately discriminate between alleles:
  • Exceptional differentiation of heterozygous and homozygous alleles
  • Discrete clusters in SNP genotyping assays

Achieve excellent reproducibility and efficiency:
  • Fast, high performance five-colored multiplex PCR
  • Similar abundance levels achieved with low copy number
  • Broad dynamic range of up to 10 orders of magnitude

Increase flexibility:
  • Compatible with both standard and fast cycling protocolsRetain accurate quantification

Quick Notes:
  • These kits are compatible with all fluorogenic probebased technologies, including hybridization probes (e.g., FRET), hydrolysis probes (e.g., TaqMan®) and displacement probes (e.g., molecular beacons).
  • Initial denaturation for 20 sec at 95ºC is sufficient for enzyme reactivation; however, optimal denaturation of complex targets may require up to 3 min denaturation.
  • For 2-step cycling, use 30 sec combined annealing/ extension/data acquisition.
  • For 3-step cycling, use 20 sec for primer annealing and 1 sec for extension/data acquisition.
  • Do not exceed 25 μL volumes.

Quality

KAPA PROBE FAST qPCR Master Mix (2X) is subjected to stringent quality control tests, is free of contaminating exoand endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

ROX/fluorescein reference dyes (depending on kit configuration) are light sensitive. Exposure to direct light for an extended period of time will result in loss of fluorescent signal intensity. Avoid repeated freezing and thawing. Always ensure that the product has been fully thawed and mixed before use.

KAPA PROBE FAST qPCR Master Mix (2X) is stable through 30 freeze-thaw cycles. Ensure that all reagents are stored protected from light at -20°C when not in use. When protected from light, reagents are stable in the dark at 4°C for at least one week, and may be stored at this temperature for short-term use provided that they do not become contaminated with microbes and/or nucleases.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

Applied Biosystems is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
QuantStudio is a trademark of IROA Technologies LLC
ROX is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Kit Components Only

Product No.
Description

  • KAPA PROBE FAST qPCR Master Mix (2X) with ROX Low

  • KAPA Taq HotStart® DNA Polymerase

  • reaction buffer

  • dNTPs

  • MgCl2 5 nM

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Henriksson et al.
Cell, 176(4), 882-896 (2019-01-15)
T helper type 2 (Th2) cells are important regulators of mammalian adaptive immunity and have relevance for infection, autoimmunity, and tumor immunology. Using a newly developed, genome-wide retroviral CRISPR knockout (KO) library, combined with RNA-seq, ATAC-seq, and ChIP-seq, we have dissected
Sydney Lavoie et al.
Gastroenterology, 158(5), 1359-1372 (2020-01-10)
Intestinal microbes and their metabolites affect the development of colorectal cancer (CRC). Short-chain fatty acids are metabolites generated by intestinal microbes from dietary fiber. We investigated the mechanisms by which free fatty acid receptor 2 (FFAR2), a receptor for short-chain

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service