KK1006
Roche
KAPA Taq ReadyMix
2 ×, suitable for PCR
Synonym(s):
PCR, taq
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106300
NACRES:
NA.54
Recommended Products
usage
50 μL sufficient for 250 reactions
Quality Level
shelf life
≤18 mo.
feature
dNTPs included
hotstart: no
packaging
pkg of 6.25 mL
manufacturer/tradename
Roche
concentration
2 ×
technique(s)
PCR: suitable
input
purified DNA
storage temp.
−20°C
General description
KAPA Taq ReadyMix™ (2X) is a ready-to-use cocktail containing all components for polymerase chain reaction (PCR), except primers and template. KAPA Taq ReadyMix is available with and without dye. The 2X ReadyMix with dye contains two inert tracking dyes to enable direct loading of PCR products onto agarose gels for analysis by electrophoresis, without the need to add a DNA loading solution. KAPA Taq DNA Polymerase is based on the single-subunit, wild-type Taq DNA polymerase of the thermophilic bacterium Thermus aquaticus.
Application
KAPA Taq ReadyMix™ has been used in:
- high throughput PCR
- amplification of low copy DNA templates
- multiplex PCR
- specific amplification of complex templates
- RT-PCR
- as a component of the reaction mixture for polymerase chain reaction (PCR) and for cDNA amplification
Biochem/physiol Actions
KAPA Taq and KAPA Taq HotStart® DNA Polymerase elicit 5′-3′ polymerase and 5′-3′ exonuclease activities, but no 3′-5′ exonuclease (proofreading) activity. The enzymes exhibit an error rate of approximately 1 error per 2.2 X 105 nucleotides incorporated. In hot start formulation, the KAPA Taq associates with a proprietary antibody that inactivates the enzyme until the first denaturation step, eliminating spurious amplification products and increasing reaction efficiency and sensitivity. PCR products generated with KAPA Taq are A-tailed and are suitable for cloning into TA cloning vectors.
Features and Benefits
High performance:
Quick Notes:
- Improved sensitivity, specificity, and yields
- Novel buffer formulation facilitates specific primer annealing, leading to higher yield of specific product
Quick Notes:
- KAPA Taq ReadyMix can replace any commercial Taq DNA polymerase in an existing protocol. The annealing temperature may need to be optimized to account for differences in formulation.
- The KAPA Taq PCR system is suitable for the amplification of fragments up to 3.5 kb from genomic DNA or 5 kb from less complex targets.
- The 2X KAPA Taq ReadyMix with dye includes two inert tracking dyes, which allow loading of PCR products directly onto agarose gels for analysis.
- KAPA Taq ReadyMixes contain 1.5 mM MgCl2 and 0.2 mM of each dNTP (at 1X).
Quality
Each batch of KAPA Taq DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA Taq ReadyMixes are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activity, and meet strict requirements with respect to DNA contamination levels.
Preparation Note
Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for longterm storage
Other Notes
For Research Use Only. Not for use in diagnostic procedures.
Legal Information
HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
Kit Components Only
Product No.
Description
- KAPA Taq Standard or HotStart® DNA Polymerase 5 U/μL
- 10X KAPA Taq Buffer A
- 10X KAPA Taq Buffer B
- 10X KAPA Buffer
- 5X KAPA Taq HotStart® Buffer (HotStart® kits only)
- MgCl2 25 mM
- dNTP Mix (optional) 10 mM each
- Stabilizers
See All (8)
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Expanding the host range of small insect RNA viruses: Providence virus (Carmotetraviridae) infects and replicates in a human tissue culture cell line.
Jiwaji M, et al.
The Journal of General Virology, 97(10), 2763-2768 (2016)
Meesbah Jiwaji et al.
PloS one, 14(6), e0217494-e0217494 (2019-06-05)
Emerging viral diseases, most of which are zoonotic, pose a significant threat to global health. There is a critical need to identify potential new viral pathogens and the challenge is to identify the reservoirs from which these viruses might emerge.
W Malherbe et al.
International journal for parasitology. Parasites and wildlife, 10, 207-210 (2019-11-02)
This study reports on the first evidence of genomic material of the causative agent for epizootic ulcerative syndrome (EUS), Aphanomyces invadans, from fish in the Limpopo River system and the Kruger National Park, South Africa. Fourteen fish species were collected
Effects of inoculating Lachnum and Cadophora isolates on the growth of Vaccinium corymbosum.
Bizabani C and Dames J
Microbiological research, 181, 68-74 (2015)
Mariska R Greeff et al.
Diseases of aquatic organisms, 99(2), 103-117 (2012-06-14)
Abalone Haliotis midae exhibiting typical clinical signs of tubercle mycosis were discovered in South African culture facilities in 2006, posing a significant threat to the industry. The fungus responsible for the outbreak was identified as a Peronosporomycete, Halioticida noduliformans. Currently
Articles
An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service