9801-OP
Water
OmniPur® Grade, Sterile, Nuclease Free
Synonym(s):
Water
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
50202301
NACRES:
NA.05
bp:
100 °C (lit.)
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
clear colorless
refractive index
n20/D 1.34 (lit.)
bp
100 °C (lit.)
mp
0 °C (lit.)
density
1.000 g/mL at 3.98 °C (lit.)
shipped in
ambient
SMILES string
O
InChI
1S/H2O/h1H2
InChI key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Molecular biology grade.
Specifications:
DNase: None detected
Protease: None detected
RNase: None detected
Sterility: To pass test
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Application
- Microsyringe-based slug-flow microextraction for rapid and accurate determination of antibiotics in highly saline seawater.: Presents a novel water-based microextraction method for detecting antibiotics in seawater, crucial for environmental monitoring (Zhang et al., 2024).
- Assessment of Hydrophilicity/Hydrophobicity in Mesoporous Silica by Combining Adsorption, Liquid Intrusion, and Solid-State NMR Spectroscopy.: Examines water interactions with silica surfaces, offering insights critical for material science and catalysis research (Collados et al., 2024).
- A novel zwitterionic magnetic nanocomposite developed for non-invasive speciation analysis of inorganic chromium.: Discusses a water-based analytical method using a magnetic nanocomposite for environmental and health-related chromium detection (Faiz et al., 2024).
- Solving the retention time repeatability problem of hydrophilic interaction liquid chromatography.: Addresses challenges in water-based chromatography, improving the reliability and efficiency of analytical methods (Serafimov et al., 2024).
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
No data available
Flash Point(C)
No data available
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service