Skip to Content
Merck
All Photos(1)

Key Documents

8.00235

Sigma-Aldrich

Cinnamic acid

for synthesis

Synonym(s):

Cinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOH
CAS Number:
Molecular Weight:
148.16
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-398-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.3 hPa ( 128 °C)

Quality Level

Assay

≥99.0% (HPLC)

form

powder

potency

2500 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

3-4 (20 °C, 0.4 g/L in H2O)

bp

300 °C/1013 hPa

mp

132-136 °C

transition temp

flash point 160 °C

solubility

0.4 g/L

density

1.25 g/cm3 at 4 °C

bulk density

500 kg/m3

storage temp.

no temp limit

SMILES string

OC(=O)\C=C\c1ccccc1

InChI

1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+

InChI key

WBYWAXJHAXSJNI-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Biodegradable polymer enhancements: A study investigated the biodegradability of chitosan films modified with cinnamic acid and ellagic acid, highlighting improvements in environmental degradation rates, which could be beneficial in packaging and agricultural applications (Michalska-Sionkowska et al., 2024).
  • Enzymatic studies in alkaloid biosynthesis: Research on the enzymes cinnamate 4-hydroxylase (CYP73A) and p-coumaroyl 3′-hydroxylase (CYP98A) from Leucojum aestivum explored their roles in the biosynthesis of Amaryllidaceae alkaloids, emphasizing cinnamic acid′s importance in metabolic pathways essential for pharmaceutical applications (Desgagné-Penix et al., 2024).
  • Quality evaluation in traditional medicine: A metabolomics approach was used to assess the quality of Scrophularia ningpoensis hemsl by identifying biomarkers, including cinnamic acid derivatives, thus providing a method for quality control in herbal medicine production (Lu et al., 2024).

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 132 °C
Melting range (upper value): ≤ 136 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service