Skip to Content
Merck
All Photos(1)

Key Documents

537088

Millipore

Pronase® Protease, Streptomyces griseus, Nuclease-Free

Pronase® Protease, Streptomyces griseus, Nuclease-Free

Widely used broad-spectrum protease mixture

Synonym(s):

Nuclease-Free Protease, Streptomyces griseus, Pronase® Protease, Pronase® Protease, Streptomyces griseus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

description

RTECS - UK9595000

form

lyophilized

specific activity

≥70,000 proteolytic units/g dry wt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 10 mg/mL

foreign activity

Nuclease, none detected

shipped in

ambient

storage temp.

2-8°C

General description

PRONASE® Protease, Streptomyces griseus, nuclease-free is a non-specific protease that liquefies mucins and digests proteins to free amino acids. This mixture can be used to isolate living chondrocytes. It is most active at pH 7.0 but stable over wide ranges of pH and temperature. It has an optimal pH of 7.5. Pronase is less cytotoxic and produces more efficient dissociation than collagenase.[1]

Application

Pronase® Protease, Streptomyces griseus, nuclease-free has been used:
  • in single-cell preparations for cardiac side population (CSP) analysis[1]
  • in aflatoxin plasma biomarker assay[2]
  • to permeabilize Caenorhabditis elegans embryo for in vivo micrococcal nuclease digestion[3]

Biochem/physiol Actions

Proteases catalyze the hydrolysis of proteins to form peptides and free amino acids. They also regulate the breakdown of fats and carbohydrates through many enzymatic cascades and activating proenzyme zymogens.[4]

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will liberate a digestion product equivalent to 25 µg of tyrosine per min at 40°C, pH 7.5.

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Note: 1 KU = 1000 units.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PRONASE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ehsan Pourkarimi et al.
eLife, 5 (2016-12-24)
The primary task of developing embryos is genome replication, yet how DNA replication is integrated with the profound cellular changes that occur through development is largely unknown. Using an approach to map DNA replication at high resolution in C. elegans
Mustafa Mahfuz et al.
Journal of exposure science & environmental epidemiology, 29(5), 655-662 (2018-09-07)
Dietary exposure to aflatoxin is implicated in growth faltering of children. Despite the high burden of childhood stunting in urban Bangladesh, there are no data on long-term exposure to aflatoxin. This study aimed to explore aflatoxin exposure levels in a
Industrial Biotechnology and Commodity Products
Comprehensive Biotechnology, 3, 587-603 null
Annette Meeson et al.
Stem cell reviews and reports, 9(5), 731-739 (2013-04-27)
Cardiac side population (CSP) cells, defined by their ability to efflux the vital dye Hoechst 33342, have been identified as putative cardiac stem cells based on their potential to give rise to both cardiomyocytes and endothelial cells. The CSP phenotype

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service