Skip to Content
Merck
All Photos(1)

Key Documents

1.04716

Millipore

Imidazole

buffer substance ACS

Synonym(s):

Imidazole, Glyoxaline, Iminazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
MDL number:
UNSPSC Code:
12161703
EC Index Number:
206-019-2
NACRES:
NB.21
grade:
ACS reagent
bp:
256 °C/1013 hPa
vapor pressure:
0.003 hPa ( 20 °C)

grade

ACS reagent

Quality Level

vapor pressure

0.003 hPa ( 20 °C)

Assay

≥99% (perchloric acid titration)

form

solid

autoignition temp.

480 °C

potency

970 mg/kg LD50, oral (Rat)

dilution

(for analytical testing)

pH

10.5 (20 °C, 67 g/L in H2O)

bp

256 °C/1013 hPa

mp

90 °C

transition temp

flash point 145 °C

solubility

633 g/L

density

1.233 g/cm3 at 20 °C

bulk density

500‑600 kg/m3

storage temp.

15-25°C

SMILES string

[nH]1cncc1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 99
Identity (IR-spectrum): conforms
pH-value (5 %, in water): 9.5 - 11.0
Melting point: 90 - 92
UV/VIS Spectrum: conforms
Fe (Iron): ≤ 0.001 %
Sulfated ash: ≤ 0.1
Water (according to Karl Fischer): ≤ 0.2
Corresponts to ACS

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service