1.00246
Platinum Cobalt Color Standard Solution (HAZEN 500)
Pt 500 mg/L Certipur®
Synonym(s):
Hazen Color Standard Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
General description
Platinum Cobalt Color Standard Solution is gravimetrically prepared using potassium hexachloroplatinate and cobalt chloride hexahydrate in hydrochloric acid. It is manufactured, qualified, and distributed in accordance with DIN EN ISO 9001:2008. It is traceable to NIST SRM 930e and has been analyzed using the double aperture method by a high-performance UV/VIV/NIR spectrophotometer in the monochromatic spectral range of 190-800 nm.
Application
The color standard solution can be used for visual comparison or as a calibration standard for instrumental analysis. It can also be used to prepare color reference solutions of different Hazen units.
Features and Benefits
- Stable solution with a minimum shelf life of 3 years
- Accompanied by a detailed certificate of analysis (CoA)
- Compliant to DIN EN ISO 7887, ASTM D 1209-05, DIN EN ISO 6271, and APHA 2120 B
Legal Information
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Carc. 1B - Met. Corr. 1
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service