Skip to Content
Merck
All Photos(1)

Key Documents

V-026

Supelco

D3-Vitamin D2 (6,19,19-D3) solution

100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Vitamin D2 (6,19,19-d3) solution, Calciferol (6,19,19-d3) solution, Ercalciol (6,19,19-d3) solution, Ergocalciferol (6,19,19-d3) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28D3H41O
Molecular Weight:
399.67
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Shoot®)
(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in ethanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

mass shift

M+3

storage temp.

−20°C
−70°C

SMILES string

[2H]/C([2H])=C1CC[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)\C=C\[C@H](C)C(C)C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1/i4D2,13D

InChI key

MECHNRXZTMCUDQ-OQGZBSKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for quantitation of Vitamin D2 levels in LC-MS/MS methods and testing applications including clinical and diagnostic testing or quality control testing of vitamin supplements, fortified foods or pharmaceutical preparations. Vitamin D2, also called ergocalciferol, is a form of vitamin D sold as a vitamin D supplement under trade names such as Deltalin, Drisdol®, and Calcidol®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Calcidol is a registered trademark of Patrin Pharma, Inc.
Drisdol is a registered trademark of Sanofi-Aventis
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - (ethanol)

Flash Point(C)

12 °C - (ethanol)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service