Skip to Content
Merck
All Photos(1)

Documents

L-022

Supelco

Lamotrigine-13C, 15N4 solution

500 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC8H7Cl215N4N
Molecular Weight:
261.06
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Nc1[15n][13c]([15NH2])[15n][15n]c1-c2cccc(Cl)c2Cl

InChI

1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)/i9+1,13+1,14+1,15+1,16+1

InChI key

PYZRQGJRPPTADH-JCDNYDJVSA-N

General description

A certified solution and internal standard suitable for GC/MS or LC/MS applications such as clinical toxicology, urine drug testing, forensic analysis or isotope dilution methods. Lamotrigine is an antiepileptic drug marketed under the trade name Lamictal for the treatment of epilepsy and bipolar disorder.

Application


  • Quantitative analysis in pharmaceutical research: The use of Lamotrigine-¹³C, ¹⁵N₄ in neuropharmacological studies is highlighted by its role in enhancing the precision of quantitation methods for drugs and metabolites using LC-MS/MS. This application demonstrates the importance of stable isotope-labeled compounds in ensuring accuracy and reliability in drug metabolism studies (Li et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service