Skip to Content
Merck
All Photos(1)

Key Documents

900105P

Avanti

PGF1α

Prostaglandin F1α, Avanti Research - A Croda Brand 900105P, powder

Synonym(s):

Prostaglandin F1α

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36O5
CAS Number:
Molecular Weight:
356.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (900105P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900105P

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H36O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-19,21-23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,18-,19+/m0/s1

InChI key

DZUXGQBLFALXCR-CDIPTNKSSA-N

General description

Prostaglandins (PGs) contain prostanoic acid. They are divided as PGA, PGB, PGC, PGD, PGE and PGF groups. They all contain a five-member ring structure differing in their functions. PGF is known to be more soluble in phosphate buffer. PGF1α is a serum eicosanoid.

Biochem/physiol Actions

PGs mediate vasodilation and vasoconstriction.

Packaging

5 mL Amber Glass Screw Cap Vial (900105P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prostaglandin synthesis, 23-23 (2012)
Liang Cui et al.
PLoS neglected tropical diseases, 7(8), e2373-e2373 (2013-08-24)
Dengue virus (DENV) is the most widespread arbovirus with an estimated 100 million infections occurring every year. Endemic in the tropical and subtropical areas of the world, dengue fever/dengue hemorrhagic fever (DF/DHF) is emerging as a major public health concern.
The Prostaglandins (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service