Skip to Content
Merck
All Photos(1)

Key Documents

870719P

Avanti

18:1 (n9) Coenzyme A

Avanti Research - A Croda Brand 870719P, powder

Synonym(s):

(9Z-octadecenoyl) Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H77N10O17P3S
CAS Number:
Molecular Weight:
1083.07
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (with stopper and crimp cap (870719P-5mg))

manufacturer/tradename

Avanti Research - A Croda Brand 870719P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCC/C=C\CCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C39H68N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-30(48)67-23-22-41-29(47)20-21-42-37(51)34(50)39(2,3)25-60-66(57,58)63-65(55,56)59-24-28-33(62-64(52,53)54)32(49)38(61-28)46-27-45-31-35(40)43-26-44-36(31)46;;;/h11-12,26-28,32-34,38,49-50H,4-10,13-25H2,1-3H3,(H,41,47)(H,42,51)(H,55,56)(H,57,58)(H2,40,43,44)(H2,52,53,54);3*1H3/b12-11-;;;/t28-,32?,33+,34+,38-;;;/m1.../s1

InChI key

NKHKRXKEDWPIOC-UQNGRMJFSA-N

General description

18:1 (n9) Coenzyme A or oleoyl-CoA (18:1n-9) is a coenzyme A derivative of oleic acid. It is highly found in cutaneous lipids.

Application

18:1 (n9) Coenzyme A or (9Z-octadecenoyl) Coenzyme A (ammonium salt) has been used along with 15N apo-ACP to prepare isotopically labeled acyl-ACP standard.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (870719P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harini Sampath et al.
The Journal of biological chemistry, 289(5), 2482-2488 (2013-12-21)
The skin is the single largest organ in humans, serving as a major barrier to infection, water loss, and abrasion. The functional diversity of skin requires the synthesis of large amounts of lipids, such as triglycerides, wax esters, squalene, ceramides
Jeong-Won Nam et al.
The Plant cell, 32(4), 820-832 (2020-02-16)
Acyl carrier proteins (ACPs) are the scaffolds for fatty acid biosynthesis in living systems, rendering them essential to a comprehensive understanding of lipid metabolism. However, accurate quantitative methods to assess individual acyl-ACPs do not exist. We developed a robust method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service