Skip to Content
Merck
All Photos(1)

Key Documents

860476P

Avanti

1-desoxymethylsphinganine-d5

1-desoxymethylsphinganine-d5 (m17:0), powder

Synonym(s):

1-Deoxymethylsphinganine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H32D5NO
CAS Number:
Molecular Weight:
276.51
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860476P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860476P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](CN)([H])CCCCCCCCCCCCCC(C([2H])([2H])[2H])([2H])[2H]

InChI key

UGVBFHUWZNNKIK-RPRHKWDUSA-N

General description

Desoxymethylsphinganine is a are the major sphingoid-base present in mammals.

Application

1-desoxymethylsphinganine-d5 may be used as an internal standard for the sphingoid analysis from plasma and tissue.

Biochem/physiol Actions

Mutations in the palmitoyltransferase (SPT) in hereditary sensory neuropathy type 1 (HSAN-1), due to altered substrate specificity 1-desoxymethylsphinganine and 1-deoxy-sphinganine.

Packaging

5 mL Amber Glass Screw Cap Vial (860476P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oral L-serine supplementation reduces production of neurotoxic deoxysphingolipids in mice and humans with hereditary sensory autonomic neuropathy type 1
Garofalo K, et al.
The Journal of Clinical Investigation, 121(12), 4735-4745 (2011)
Annelies Rotthier et al.
Human mutation, 32(6), E2211-E2225 (2011-05-28)
Hereditary sensory and autonomic neuropathy type I (HSAN-I) is an axonal peripheral neuropathy leading to progressive distal sensory loss and severe ulcerations. Mutations in SPTLC1 and SPTLC2, encoding the two subunits of serine palmitoyltransferase (SPT), the enzyme catalyzing the first
Biophysical properties of novel 1-deoxy-(dihydro) ceramides occurring in mammalian cells
Jimenez-Rojo N, et al.
Biophysical Journal, 107(12), 2850-2859 (2014)
Gergely Karsai et al.
The Journal of biological chemistry, 295(7), 1889-1897 (2019-12-22)
Sphingolipids (SLs) are structurally diverse lipids that are defined by the presence of a long-chain base (LCB) backbone. Typically, LCBs contain a single Δ4E double bond (DB) (mostly d18:1), whereas the dienic LCB sphingadienine (d18:2) contains a second DB at
Hereditary sensory neuropathy type 1 is caused by the accumulation of two neurotoxic sphingolipids
Penno A, et al.
The Journal of Biological Chemistry, 285(15), 11178-11187 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service