Skip to Content
Merck
All Photos(2)

Key Documents

860383P

Avanti

18:0 PG-d70

Avanti Research - A Croda Brand 860383P, powder

Synonym(s):

1,2-distearoyl-d70-sn-glycero-3-[phospho-rac-(1-glycerol)] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H12O10PNaD70
CAS Number:
Molecular Weight:
871.49
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860383P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860383P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H]

General description

1,2-distearoyl-d70-sn-glycero-3-[phospho-rac-(1-glycerol)] (18:0 PG-d70) is a deuterated phosphoglycerol (PG) wherein 70 protons in distearoyl are replaced by deuterium.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Packaging

5 mL Amber Glass Screw Cap Vial (860383P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of materials chemistry. B (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service