850173P
Avanti
rac-16:0 PI(5)P-d5
Avanti Research™ - A Croda Brand 850173P, powder
Synonym(s):
1,2-dipalmitoyl-rac-glycero(d5)-3-phospho-(1′-myo-inositol-5′-phosphate) (ammonium salt)
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 100 μg (with stopper and crimp cap (850173P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850173P-500ug))
manufacturer/tradename
Avanti Research™ - A Croda Brand 850173P
shipped in
dry ice
storage temp.
−20°C
SMILES string
[2H]C(C([2H])([2H])OP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCCCCCCCCCC)=O)C([2H])([2H])OC(CCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]
InChI key
XAERIJXJLCXLHF-QKBLWVTASA-N
General description
Application
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service