Skip to Content
Merck
All Photos(1)

Key Documents

840526P

Avanti

Hydrogenated MGDG

Avanti Research - A Croda Brand, powder

Synonym(s):

Monogalactosyldiacylglycerol (plant, hydrogenated)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H82O10
CAS Number:
Molecular Weight:
759.11
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840526P-10MG)
pkg of 1 × 25 mg (840526P-25MG)
pkg of 1 × 5 mg (840526P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](COC(CCCCCCCCCCCCCCCCC)=O)(OC(CCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C43H82O10/c1-3-5-7-9-11-13-15-17-18-20-21-23-25-27-29-31-38(45)50-34-36(35-51-43-42(49)41(48)40(47)37(33-44)53-43)52-39(46)32-30-28-26-24-22-19-16-14-12-10-8-6-4-2/h36-37,40-44,47-49H,3-35H2,1-2H3/t36-,37-,40+,41+,42-,43-/m1/s1

InChI key

ZCXQDNWIQSAJIR-GREPGOCTSA-N

General description

Monogalactosyldiacylglycerol (MGDG), a galactosylglycerolipid, is a major lipid in the thylakoid membrane of the chloroplast of leaves. It is also localized in the roots, fruits and grains. MGDG is synthesized from Monogalactosyldiacylglycerol synthase 1 (MGD1). It is a precursor for digalactosyldiacylglycerol (DGDG).

Biochem/physiol Actions

Monogalactosyldiacylglycerol (MGDG) plays an important role in biogenesis and integrity of plastids for photosynthesis, as they need a galactolipid-rich environment.

Packaging

5 mL Amber Glass Screw Cap Vial (840526P-5MG, 840526P-10MG, and 840526P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro reconstitution of monogalactosyldiacylglycerol (MGDG) synthase regulation by thioredoxin
Yamaryo Y, et al.
Febs Letters, 580(17), 4086-4090 (2006)
The galactolipid monogalactosyldiacylglycerol (MGDG) contributes to photosynthesis-related processes in Arabidopsis thaliana
Aronsson H
Plant signaling & behavior, 3(12), 1093-1095 (2008)
Digestion of plant monogalactosyldiacylglycerol and digalactosyldiacylglycerol in rat alimentary canal
Sugawara T and Miyazawa T
The Journal of Nutritional Biochemistry, 11(3), 147-152 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service