Skip to Content
Merck
All Photos(1)

Key Documents

810257P

Avanti

C16/C11-TF/C12-DNP TG

Avanti Research - A Croda Brand 810257P, powder

Synonym(s):

1-O-hexadecyl-2-(dipyrrometheneboron difluoride)undecanoyl-3-(dinitrophenylamino)dodecanoyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H98BF2N5O9
CAS Number:
Molecular Weight:
1094.27
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810257P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810257P

shipped in

dry ice

storage temp.

−20°C

General description

Triglyceride molecules are the major form of storage and transport fatty acids present in plasma and within cells. C16/C11-TF/C12-DNP TG has the Dinitro phenyl group attached through carbon 12.

Biochem/physiol Actions

Triglycerides can cross the blood-brain barrier and are found in human cerebrospinal fluid (CSF). The metabolism of triglycerides and fatty acids takes place in the liver. Accumulation of triglycerides within the hepatocytes due to overnutrition and obesity leads to a clinical condition called as non-alcoholic fatty liver disease (NAFLD).

Packaging

5 mL Amber Glass Screw Cap Vial (810257P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Banks et al.
International journal of obesity (2005), 42(3), 391-397 (2017-10-11)
Resistance at the brain receptors for leptin and insulin has been associated with increased feeding, obesity and cognitive impairments. The causal agent for central resistance is unknown but could be derived from the blood. Here we postulate whether hypertriglyceridemia, the
Michele Alves-Bezerra et al.
Comprehensive Physiology, 8(1), 1-8 (2018-01-23)
Triglyceride molecules represent the major form of storage and transport of fatty acids within cells and in the plasma. The liver is the central organ for fatty acid metabolism. Fatty acids accrue in liver by hepatocellular uptake from the plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service