Skip to Content
Merck
All Photos(2)

Key Documents

700368P

Avanti

3α,7α-dihydroxy-5β -cholestanoic acid-d5

Avanti Research - A Croda Brand

Synonym(s):

3α,7α-dihydroxy-5β -cholestanoic acid-d5, 3α,7α-dihydroxy-5β -cholestanate-d5, 3α,7α-dihydroxycholestanoic acid-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H41D5O4
Molecular Weight:
439.68
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700368P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Application

lipidomics

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service