Key Documents
Z742596
Accuris SmartDoc narrow band pass filter
590nm filter; Used for imaging EtBR
Synonym(s):
Accuris SmartDoc
About This Item
UNSPSC Code:
41106000
NACRES:
NB.24
feature
590nm filter; Used for imaging EtBR
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific E5001-590
General description
A narrow band pass filter of 590 nm to use for imaging EtBR on a transilluminator
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.
Visit the Document Library
Customers Also Viewed
Slide 1 of 1