Skip to Content
Merck
All Photos(1)

Key Documents

Z660655

Plas-Labs palladium canister

used with the Plas-Labs catalyst heaters

Synonym(s):

humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103406
NACRES:
NB.35

manufacturer/tradename

Plas-Labs

General description

This replacement Plas-Labs palladium canister is used with the Plas-Labs catalyst heaters to regulate the internal atmosphere for the Plas-Labs temperature and humidity boxes. Any oxygen present will react with an anaerobic gas mixture and be reduced to water vapor.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service