Z511501
Kimax® heavy-duty wide-mouth, large numbers volumetric flasks - CLASS A
capacity 10 mL
Synonym(s):
92812G-10
About This Item
Recommended Products
material
glass
wide-mouth
Joint
joint ST/NS neck6
joint stopper6
sterility
non-sterile
feature
class A USP Type I and ASTM E438, Type I
closure type Glass stopper
packaging
pack of 1
case of 6
manufacturer/tradename
Kimble® 92812G-10
tol. measuring range
0.03 mL
capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
Supplied with glass stopper.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service