material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
Cowie 005.540.1
L
400 mm
shaft diam.
6 mm
Related Categories
General description
With 6.5mm diamond-shaped button and steel core, order blades below.
Stirring shaft with steel core for optimal rigidity. Use with PTFE stirrer guides and for applications which do not require a guide. Unbreakable, scratch resistant, and usable up to 280°C.